This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00627014_g1
See other EDNRB GE Assays ›
Gene Symbol:
Ednrb
Gene Name:
endothelin receptor type B
Gene Aliases:
ET-B, ET-BR, ETR-b, ETb, Sox10m1
Chromosome Location:
Chr.14: 103814615 - 103844476 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00627014_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ednrb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136061.2 NP_001129533.1 3 - 4 1007 129
NM_001276296.1 NP_001263225.1 2 - 3 821 129
NM_007904.4 NP_031930.1 2 - 3 821 129
GenBank mRNA AK076426.1 - 2 - 3 815 129
AK082103.1 - 3 - 4 808 129
AK083415.1 - 3 - 4 704 129
AK085165.1 - 2 - 3 812 129
AK085532.1 - 2 - 3 817 129
AK195461.1 - 2 - 3 270 129
AK198739.1 - 2 - 3 254 129
AK199447.1 - 2 - 3 246 129
AK202909.1 - 2 - 3 218 129
AK204434.1 - 2 - 3 240 129
AK211790.1 - 2 - 3 281 129
AK215891.1 - 2 - 3 147 129
BC026553.1 - 3 - 4 933 4971909 129
U32329.1 - 2 - 3 799 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002641.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ednrb

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
positive regulation of protein phosphorylation
regulation of pH
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
peripheral nervous system development
posterior midgut development
regulation of blood pressure
positive regulation of cell proliferation
negative regulation of neuron maturation
vein smooth muscle contraction
sensory perception of pain
cGMP-mediated signaling
melanocyte differentiation
regulation of fever generation
negative regulation of cellular protein metabolic process
response to lipopolysaccharide
enteric smooth muscle cell differentiation
positive regulation of urine volume
positive regulation of renal sodium excretion
epithelial fluid transport
vasoconstriction
vasodilation
negative regulation of apoptotic process
pigmentation
developmental pigmentation
macrophage chemotaxis
response to pain
enteric nervous system development
regulation of epithelial cell proliferation
regulation of sensory perception of pain
positive regulation of penile erection
endothelin receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity
endothelin receptor activity
protein binding
peptide hormone binding
type 1 angiotensin receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019783.mm.1 NM_001136061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019783.mm.1 NM_001276296.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019783.mm.1 NM_007904.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400019783.mm.1 TC1400002641.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0