This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00626465_m1
See other ARHGAP33 GE Assays ›
Gene Symbol:
Arhgap33
Gene Name:
Rho GTPase activating protein 33
Gene Aliases:
AW324378, NOMA-GAP, Snx26, TCGAP
Chromosome Location:
Chr.7: 30522226 - 30535092 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00626465_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289670.1 NP_001276599.1 15 - 16 134
NM_001289682.1 NP_001276611.1 16 - 17 134
NM_178252.3 NP_839983.1 16 - 17 134
GenBank mRNA AK142180.1 - 15 - 16 134
AY217764.1 - 15 - 16 134
BC061471.1 - 9 - 10 5698780 134
BC065086.1 - 16 - 17 6850475 134
BC065166.1 - 16 - 17 6836510 134
BC066047.1 - 16 - 17 6822191 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002763.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap33

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

transport
signal transduction
protein transport
positive regulation of GTPase activity
GTPase activator activity
protein binding
phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700024537.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024537.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024537.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024537.mm.1 TC0700002763.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0