This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624973_m1
See other ADGRD1 GE Assays ›
Gene Symbol:
Adgrd1
Gene Name:
adhesion G protein-coupled receptor D1
Gene Aliases:
E230012M21Rik, Gpr133
Chromosome Location:
Chr.5: 129096666 - 129204599 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624973_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081342.1 NP_001074811.1 22 - 23 2760 110
GenBank mRNA AK054021.1 - 7 - 8 607 110
AK153762.1 - 22 - 23 2763 110
BC151161.1 - 22 - 23 2462 9088050 110
BC157994.1 - 21 - 22 2366 9007307 110
BC158001.1 - 21 - 22 2366 9007310 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001501.mm.1

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013520.mm.1 NM_001081342.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013520.mm.1 NM_001081342.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500013520.mm.1 TC0500001501.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0