This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624670_m1
See other VWA3A GE Assays ›
Gene Symbol:
Vwa3a
Gene Name:
von Willebrand factor A domain containing 3A
Gene Aliases:
E030013G06Rik
Chromosome Location:
Chr.7: 120735693 - 120805546 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624670_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vwa3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177697.3 NP_808365.2 20 - 21 2456 98
GenBank mRNA AK086942.1 - 19 - 20 2054 98
AK132732.1 - 21 - 22 3089 98
AK136898.1 - 19 - 20 2055 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001734.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vwa3a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700014682.mm.1 NM_177697.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014682.mm.1 NM_177697.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700014682.mm.1 TC0700001734.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0