This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624272_m1
See other ADGRA1 GE Assays ›
Gene Symbol:
Adgra1
Gene Name:
adhesion G protein-coupled receptor A1
Gene Aliases:
2900059M17Rik, D7Ertd680e, Gpr123
Chromosome Location:
Chr.7: 139834084 - 139878088 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624272_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgra1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177469.3 NP_803420.2 3 - 4 1139 76
GenBank mRNA AB096262.1 - 1 - 2 202 76
AK034188.1 - 2 - 3 702 76
AK082207.1 - 3 - 4 1018 76
BC056493.1 - 1 - 2 423 5704932 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK034188.1, AK082207.1, BC056493.1, NM_177469.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgra1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700017503.mm.1 NM_177469.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017503.mm.1 NM_177469.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700017503.mm.1 TC0700001996.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700017502.mm.1 NM_177469.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017502.mm.1 NM_177469.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700017502.mm.1 TC0700001996.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0