This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00619072_g1
See other SON GE Assays ›
Gene Symbol:
Son
Gene Name:
Son DNA binding protein
Gene Aliases:
2900011L12Rik, AA409051, AU067731, C81487, mKIAA1019, nrebp
Chromosome Location:
Chr.16: 91647824 - 91679195 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00619072_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Son
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178880.4 NP_849211.3 11 - 12 7354 61
GenBank mRNA AK035150.1 - 4 - 5 2845 61
AK122418.1 - 10 - 11 4470 61
BC145121.1 - 11 - 12 7294 61
CB849818.2 - 4 - 5 452 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001008.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Son

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
cytokinesis
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
cell cycle
RNA splicing
negative regulation of apoptotic process
regulation of RNA splicing
regulation of cell cycle
nucleic acid binding
DNA binding
RNA binding
protein binding
poly(A) RNA binding
RS domain binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600007854.mm.1 NM_178880.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007854.mm.1 NM_178880.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600007854.mm.1 TC1600001008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007851.mm.1 NM_178880.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007851.mm.1 NM_178880.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600007851.mm.1 TC1600001008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007852.mm.1 NM_178880.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007852.mm.1 NM_178880.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600007852.mm.1 TC1600001008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007853.mm.1 NM_178880.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007853.mm.1 NM_178880.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600007853.mm.1 TC1600001008.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 429618 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0