This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00618293_m1
See other SCCPDH GE Assays ›
Gene Symbol:
Sccpdh
Gene Name:
saccharopine dehydrogenase (putative)
Gene Aliases:
AW214504, C330023F11Rik
Chromosome Location:
Chr.1: 179668231 - 179687184 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00618293_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sccpdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178653.3 NP_848768.1 7 - 8 932 77
GenBank mRNA AK042919.1 - 7 - 8 820 77
AK046033.1 - 7 - 8 933 77
AK046468.1 - 7 - 8 920 77
AK050554.1 - 7 - 8 881 77
AK077695.1 - 7 - 8 882 77
AK082803.1 - 7 - 8 817 77
AK086448.1 - 7 - 8 955 77
AK086632.1 - 7 - 8 930 77
AK166018.1 - 7 - 8 844 77
AK212321.1 - 1 - 2 82 77
BC025803.1 - 7 - 8 929 5100552 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001699.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sccpdh

Gene Ontology Categories:

Function(s) Process(es)

biological_process
oxidation-reduction process
molecular_function
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100014050.mm.1 NM_178653.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100014050.mm.1 NM_178653.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100014050.mm.1 TC0100001699.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0