This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00617632_g1
See other CDC23 GE Assays ›
Gene Symbol:
Cdc23
Gene Name:
CDC23 cell division cycle 23
Gene Aliases:
6030435O18, D18Ertd243e
Chromosome Location:
Chr.18: 34630946 - 34651745 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00617632_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdc23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178347.4 NP_848124.1 7 - 8 866 81
GenBank mRNA AK031459.1 - 7 - 8 849 81
AK038523.1 - 7 - 8 849 81
AK047789.1 - 7 - 8 849 81
AK048246.1 - 7 - 8 849 81
AK052567.1 - 7 - 8 866 81
AK075948.1 - 7 - 8 867 81
AK081461.1 - 7 - 8 867 81
AK169677.1 - 3 - 4 340 81
BC059013.1 - 7 - 8 835 6813730 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001154.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdc23

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
mitotic metaphase plate congression
regulation of mitotic metaphase/anaphase transition
cell division
protein K11-linked ubiquitination
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800008480.mm.1 NM_178347.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008480.mm.1 NM_178347.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800008480.mm.1 TC1800001154.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0