Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035240.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035240.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035241.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035241.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035242.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035242.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035243.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_001035243.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_177341.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
NM_177341.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003532.mm.1 |
TC1900000356.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035239.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035239.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035240.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035240.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035241.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035241.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035242.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035242.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035243.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_001035243.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_177341.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
NM_177341.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900003531.mm.1 |
TC1900000356.mm.1 |
MTA TC |