This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00616432_g1
See other EXOC3 GE Assays ›
Gene Symbol:
Exoc3
Gene Name:
exocyst complex component 3
Gene Aliases:
2810050O03Rik, E430013E20Rik, Sec6, Sec6l1
Chromosome Location:
Chr.13: 74169796 - 74208729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00616432_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Exoc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177333.3 NP_796307.2 8 - 9 1592 121
GenBank mRNA AK088333.1 - 8 - 9 1586 121
AK089979.1 - 8 - 9 1602 121
AK131141.1 - 8 - 9 1576 121
AK141537.1 - 8 - 9 1592 121
BC031743.1 - 8 - 9 1580 4187527 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002313.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Exoc3

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
exocytosis
biological_process
protein transport
exocyst localization
SNARE binding
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300015189.mm.1 NM_177333.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300015189.mm.1 NM_177333.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300015189.mm.1 TC1300002313.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0