This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00614388_m1
See other PHACTR4 GE Assays ›
Gene Symbol:
Phactr4
Gene Name:
phosphatase and actin regulator 4
Gene Aliases:
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120
Chromosome Location:
Chr.4: 132355922 - 132422446 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00614388_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phactr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161797.1 NP_001155269.1 10 - 11 1917 60
NM_175306.4 NP_780515.2 11 - 12 1998 60
GenBank mRNA AK049209.1 - 11 - 12 1961 60
AK220496.1 - 10 - 11 1896 60
BC037059.1 - 1 - 2 84 60
BC075672.1 - 11 - 12 1960 30549496 60
BC096033.1 - 11 - 12 1917 6854767 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003670.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phactr4

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
neural tube closure
Rho protein signal transduction
multicellular organism development
nervous system development
actin cytoskeleton organization
positive regulation of catalytic activity
enteric nervous system development
regulation of cell cycle
closure of optic fissure
negative regulation of integrin-mediated signaling pathway
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity
protein phosphatase type 1 activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400030097.mm.1 NM_001161797.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030097.mm.1 NM_001161797.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030097.mm.1 NM_175306.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030097.mm.1 NM_175306.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030097.mm.1 TC0400003670.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0