This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00613357_g1
See other PTK7 GE Assays ›
Gene Symbol:
Ptk7
Gene Name:
PTK7 protein tyrosine kinase 7
Gene Aliases:
8430404F20Rik, chz, mPTK7/CCK4
Chromosome Location:
Chr.17: 46564451 - 46630049 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00613357_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptk7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175168.4 NP_780377.1 12 - 13 2081 62
GenBank mRNA AF531873.1 - 12 - 13 1931 62
AK053044.1 - 12 - 13 2014 62
AK081793.1 - 2 - 3 2228 62
AK163135.1 - 12 - 13 2014 62
BC027800.1 - 1 - 2 136 62
BC076578.1 - 12 - 13 2081 30619280 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002181.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptk7

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

establishment of planar polarity
neural tube closure
axis elongation
protein phosphorylation
cell adhesion
positive regulation of neuron projection development
Wnt signaling pathway
cell migration
actin cytoskeleton reorganization
wound healing
establishment of epithelial cell apical/basal polarity
convergent extension
canonical Wnt signaling pathway
lung-associated mesenchyme development
cellular response to retinoic acid
cochlea morphogenesis
planar cell polarity pathway involved in neural tube closure
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700020564.mm.1 NM_175168.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700020564.mm.1 NM_175168.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700020564.mm.1 TC1700002181.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0