This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00613002_m1
See other EMC3 GE Assays ›
Gene Symbol:
Emc3
Gene Name:
ER membrane protein complex subunit 3
Gene Aliases:
0610039A15Rik, AI225901, AW260416, Pob, Tmem111
Chromosome Location:
Chr.6: 113514887 - 113531638 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00613002_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Emc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175101.3 NP_780310.1 1 - 2 304 65
GenBank mRNA AK020058.1 - 1 - 2 275 65
AK075595.1 - 1 - 2 305 65
AK167819.1 - 1 - 2 153 65
AK206599.1 - 1 - 2 50 65
AK213319.1 - 1 - 2 50 65
AK218418.1 - 1 - 2 50 65
BC004641.1 - 1 - 2 300 3157147 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002924.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Emc3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein folding in endoplasmic reticulum
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600022480.mm.1 NM_175101.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022480.mm.1 NM_175101.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600022480.mm.1 TC0600002924.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022478.mm.1 NM_175101.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022478.mm.1 NM_175101.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600022478.mm.1 TC0600002924.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0