This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00612789_m1
See other CHERP GE Assays ›
Gene Symbol:
Cherp
Gene Name:
calcium homeostasis endoplasmic reticulum protein
Gene Aliases:
5730408I11Rik, D8Wsu96e, DAN16, Scaf6
Chromosome Location:
Chr.8: 72460483 - 72476675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00612789_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cherp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138585.3 NP_613051.3 9 - 10 1483 76
GenBank mRNA AF536541.1 - 9 - 10 1385 76
AK049765.1 - 9 - 10 1393 76
AK144328.1 - 9 - 10 1403 77
AK171907.1 - 2 - 3 459 76
BC032191.1 - 9 - 10 1401 5322208 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002463.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cherp

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
cellular calcium ion homeostasis
negative regulation of cell proliferation
release of sequestered calcium ion into cytosol
positive regulation of NFAT protein import into nucleus
nucleic acid binding
RNA binding
ion channel binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800018857.mm.1 NM_138585.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800018857.mm.1 NM_138585.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800018857.mm.1 TC0800002463.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0