This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00612560_g1
See other UBA7 GE Assays ›
Gene Symbol:
Uba7
Gene Name:
ubiquitin-like modifier activating enzyme 7
Gene Aliases:
1300004C08Rik, Ube1l
Chromosome Location:
Chr.9: 107975567 - 107984056 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00612560_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uba7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023738.4 NP_076227.1 19 - 20 2450 63
GenBank mRNA AK004894.2 - 19 - 20 2451 63
AK054410.1 - 18 - 19 2542 63
AK089235.1 - 18 - 19 2343 63
AK129009.1 - 19 - 20 2360 63
AK170421.1 - 18 - 19 2528 63
AK170644.1 - 17 - 18 2909 63
AK172467.1 - 18 - 19 2788 63
BC138843.1 - 19 - 20 2367 8861865 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003335.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uba7

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein ubiquitination
modification-dependent protein catabolic process
ISG15-protein conjugation
ubiquitin activating enzyme activity
ubiquitin-protein transferase activity
ISG15 activating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900011012.mm.1 NM_023738.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011012.mm.1 NM_023738.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900011012.mm.1 TC0900003335.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0