This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00600726_g1
See other DIS3L2 GE Assays ›
Gene Symbol:
Dis3l2
Gene Name:
DIS3 mitotic control homolog (S. cerevisiae)-like 2
Gene Aliases:
4930429A22Rik, 8030493P09Rik
Chromosome Location:
Chr.1: 86703779 - 87050097 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00600726_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dis3l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172157.1 NP_001165628.1 2 - 3 221 74
NM_153530.2 NP_705758.1 2 - 3 221 74
GenBank mRNA AK029652.1 - 2 - 3 198 74
AK031180.1 - 2 - 3 206 74
AK040613.1 - 2 - 3 198 74
AK050762.1 - 2 - 3 222 74
BB654969.1 - 2 - 3 222 74
BC036177.1 - 2 - 3 182 5004341 74
CJ063044.1 - 2 - 3 200 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000794.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK029652.1,AK031180.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dis3l2

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, exonucleolytic
rRNA processing
mRNA catabolic process
cell cycle
mitotic nuclear division
negative regulation of cell proliferation
miRNA catabolic process
rRNA catabolic process
stem cell population maintenance
nuclear-transcribed mRNA catabolic process, exonucleolytic, 3'-5'
cell division
mitotic sister chromatid separation
RNA phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
ribonuclease activity
poly(U) RNA binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100006674.mm.1 NM_001172157.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006674.mm.1 NM_001172157.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006674.mm.1 NM_153530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006674.mm.1 NM_153530.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100006674.mm.1 TC0100000794.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0