This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00600239_g1
See other FKBP3 GE Assays ›
Gene Symbol:
Fkbp3
Gene Name:
FK506 binding protein 3
Gene Aliases:
25kDa, FKBP-3, FKBP25
Chromosome Location:
Chr.12: 65062432 - 65073938 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00600239_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fkbp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013902.4 NP_038930.1 2 - 3 223 133
GenBank mRNA AF110812.1 - 2 - 3 220 133
AF135595.1 - 2 - 3 225 133
AK012346.1 - 2 - 3 218 133
AK139213.1 - 1 - 2 1187 133
AK145648.1 - 2 - 3 231 133
AK150802.1 - 2 - 3 233 133
AK168641.1 - 2 - 3 230 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001832.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fkbp3

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
protein binding
FK506 binding
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200012497.mm.1 NM_013902.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012497.mm.1 NM_013902.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200012497.mm.1 TC1200001832.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0