This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00556658_m1
See other CD163L1 GE Assays ›
Gene Symbol:
Cd163l1
Gene Name:
CD163 molecule-like 1
Gene Aliases:
B430307C05, E430002D04Rik, Scart1
Chromosome Location:
Chr.7: 140197218 - 140231145 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00556658_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd163l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172909.4 NP_766497.3 2 - 3 693 118
GenBank mRNA AF539744.1 - 2 - 3 449 118
AK046675.1 - 2 - 3 694 118
AK088010.1 - 2 - 3 680 118
BC145639.1 - 2 - 3 419 118
BC145640.1 - 2 - 3 419 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002007.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd163l1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700017624.mm.1 NM_172909.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700017624.mm.1 NM_172909.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700017624.mm.1 TC0700002007.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0