This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00556496_m1
See other SLC13A4 GE Assays ›
Gene Symbol:
Slc13a4
Gene Name:
solute carrier family 13 (sodium/sulfate symporters), member 4
Gene Aliases:
9630060C05Rik, SUT-1, SUT1
Chromosome Location:
Chr.6: 35267953 - 35308126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00556496_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc13a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172892.3 NP_766480.1 3 - 4 1039 71
GenBank mRNA AK036354.1 - 3 - 4 1041 71
AK038937.1 - 3 - 4 1044 71
AK139428.1 - 3 - 4 1042 71
BC089161.1 - 3 - 4 1028 30440173 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002066.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc13a4

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600016276.mm.1 NM_172892.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016276.mm.1 NM_172892.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600016276.mm.1 TC0600002066.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0