Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500029758.mm.1 |
NM_172885.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500029758.mm.1 |
NM_172885.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500029758.mm.1 |
TC0500003286.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500029760.mm.1 |
NM_172885.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500029760.mm.1 |
NM_172885.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500029760.mm.1 |
TC0500003286.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500029759.mm.1 |
NM_172885.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500029759.mm.1 |
NM_172885.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500029759.mm.1 |
TC0500003286.mm.1 |
MTA TC |