This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00556331_m1
See other PODN GE Assays ›
Gene Symbol:
Podn
Gene Name:
podocan
Gene Aliases:
9430070G18, Pcan, SLRR5A
Chromosome Location:
Chr.4: 108014793 - 108032090 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00556331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Podn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285956.1 NP_001272885.1 6 - 7 958 90
NM_001285958.1 NP_001272887.1 6 - 7 854 90
NM_172874.3 NP_766462.2 6 - 7 880 90
GenBank mRNA AK034987.1 - 6 - 7 745 90
AK161018.1 - 6 - 7 744 90
AY313606.1 - 6 - 7 880 90
BC052487.1 - 6 - 7 704 90
BC094340.1 - 6 - 7 958 30609151 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004216.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Podn

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell proliferation
negative regulation of cell migration
collagen binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400025692.mm.1 NM_001285956.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400025692.mm.1 NM_001285958.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400025692.mm.1 NM_172874.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400025692.mm.1 TC0400004216.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0