This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555769_m1
See other ENOX1 GE Assays ›
Gene Symbol:
Enox1
Gene Name:
ecto-NOX disulfide-thiol exchanger 1
Gene Aliases:
B230207J08, D230005D02Rik
Chromosome Location:
Chr.14: 77154001 - 77721763 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555769_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Enox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253759.1 NP_001240688.1 8 - 9 1129 71
NM_172813.3 NP_766401.1 8 - 9 1129 71
GenBank mRNA AK045500.1 - 8 - 9 1047 71
AK051821.1 - 8 - 9 1077 71
AK082388.1 - 8 - 9 1130 71
AK162001.1 - 8 - 9 1078 71
BC052062.1 - 5 - 6 640 5703279 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001078.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Enox1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
rhythmic process
oxidation-reduction process
nucleotide binding
molecular_function
nucleic acid binding
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400008677.mm.1 NM_001253759.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400008677.mm.1 NM_001253759.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400008677.mm.1 NM_172813.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400008677.mm.1 NM_172813.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400008677.mm.1 TC1400001078.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0