This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555744_m1
See other GUCY1B2 GE Assays ›
Gene Symbol:
Gucy1b2
Gene Name:
guanylate cyclase 1, soluble, beta 2
Gene Aliases:
B930025B03, GC-SB2
Chromosome Location:
Chr.14: 62373023 - 62457562 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555744_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gucy1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204340.1 NP_001191269.1 12 - 13 1829 81
NM_172810.3 NP_766398.1 12 - 13 1874 81
GenBank mRNA AK047129.1 - 12 - 13 1875 81
AY236513.1 - 10 - 11 1548 81
BC137868.1 - 12 - 13 1826 8860887 81
BC144988.1 - 12 - 13 1823 9053536 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gucy1b2

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Process(es)

guanylate cyclase activity
ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0