This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555438_m1
See other DDX26B GE Assays ›
Gene Symbol:
Ddx26b
Gene Name:
DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B
Gene Aliases:
4930535D10Rik, 6330505F04Rik, D130066O12
Chromosome Location:
Chr.X: 56454821 - 56507843 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555438_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx26b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172779.4 NP_766367.1 13 - 14 2137 90
GenBank mRNA AK083942.1 - 13 - 14 2099 90
AK144026.1 - 3 - 4 1982 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000519.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx26b

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00003410.mm.1 NM_172779.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00003410.mm.1 NM_172779.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00003410.mm.1 TC0X00000519.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0