This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00555335_m1
See other FAM63B GE Assays ›
Gene Symbol:
Fam63b
Gene Name:
family with sequence similarity 63, member B
Gene Aliases:
AI465301, B230380D07Rik, mKIAA1164
Chromosome Location:
Chr.9: 70599014 - 70657296 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00555335_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam63b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172772.2 NP_766360.2 7 - 8 1604 95
GenBank mRNA AK035930.1 - 7 - 8 1605 95
AK046403.1 - 7 - 8 1591 95
AK077470.1 - 4 - 5 536 95
AK216605.1 - 2 - 3 272 95
AK220158.1 - 7 - 8 1633 95
BC049357.1 - 7 - 8 1581 95
BC055816.1 - 7 - 8 1332 95
BC058683.1 - 7 - 8 1562 6834871 95
BC069845.1 - 7 - 8 1317 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002599.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam63b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900021612.mm.1 NM_172772.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900021612.mm.1 NM_172772.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900021612.mm.1 TC0900002599.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0