This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00554864_g1
See other GCN1L1 GE Assays ›
Gene Symbol:
Gcn1l1
Gene Name:
GCN1 general control of amino-acid synthesis 1-like 1 (yeast)
Gene Aliases:
4932409G22, AL022764, G431004K08Rik, GCN1L, mKIAA0219
Chromosome Location:
Chr.5: 115565219 - 115622654 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00554864_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcn1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172719.2 NP_766307.2 47 - 48 6277 62
GenBank mRNA AB093217.1 - 23 - 24 3470 62
AK077013.1 - 11 - 12 1599 62
AK090137.1 - 47 - 48 6274 62
AK147257.1 - 47 - 48 6270 62
BC056933.1 - 13 - 14 1909 62
BC068244.1 - 3 - 4 531 62
BC150735.1 - 47 - 48 6280 9087646 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001267.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcn1l1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500011035.mm.1 NM_172719.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500011035.mm.1 NM_172719.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500011035.mm.1 TC0500001267.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0