This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00554573_m1
See other MEGF9 GE Assays ›
Gene Symbol:
Megf9
Gene Name:
multiple EGF-like-domains 9
Gene Aliases:
4933405H16Rik, 9630025C07, Egfl5, mKIAA0818
Chromosome Location:
Chr.4: 70431927 - 70534928 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00554573_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Megf9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172694.2 NP_766282.1 3 - 4 1043 73
GenBank mRNA AK035987.1 - 3 - 4 1044 73
AK039319.1 - 3 - 4 1044 73
AK122378.1 - 1034 73
BC088994.1 - 3 - 4 952 6734479 73
BC138128.1 - 3 - 4 982 8861147 73
BC138129.1 - 3 - 4 982 8861148 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002849.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Megf9

Panther Classification:

Molecular Function -

extracellular matrix protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400023310.mm.1 NM_172694.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400023310.mm.1 NM_172694.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400023310.mm.1 TC0400002849.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0