This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00554552_g1
See other GBA2 GE Assays ›
Gene Symbol:
Gba2
Gene Name:
glucosidase beta 2
Gene Aliases:
F630034E04
Chromosome Location:
Chr.4: 43566928 - 43578864 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00554552_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gba2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172692.3 NP_766280.2 6 - 7 1618 77
GenBank mRNA AK089192.1 - 6 - 7 1304 77
AK158174.1 - 6 - 7 1618 77
AK173213.1 - 6 - 7 1604 77
BC056935.1 - 6 - 7 1141 5703478 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gba2

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
glucosylceramide catabolic process
metabolic process
bile acid metabolic process
glycoside catabolic process
central nervous system neuron development
catalytic activity
glucosylceramidase activity
beta-glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0