This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00554474_m1
See other POGZ GE Assays ›
Gene Symbol:
Pogz
Gene Name:
pogo transposable element with ZNF domain
Gene Aliases:
9530006B08Rik
Chromosome Location:
Chr.3: 94823043 - 94883569 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00554474_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pogz
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165948.1 NP_001159420.1 3 - 4 462 105
NM_172683.3 NP_766271.2 3 - 4 462 105
GenBank mRNA AK035255.1 - 3 - 4 317 105
AK086062.1 - 3 - 4 318 106
AK161889.1 - 3 - 4 317 105
BC065134.1 - 3 - 4 451 105
BC066108.1 - 3 - 4 451 105
BC085188.1 - 3 - 4 322 30682742 105
BC096492.1 - 2 - 3 305 30745658 105
BC139432.1 - 3 - 4 317 8862454 105
BC139433.1 - 3 - 4 317 8862455 105
CF743949.1 - 3 - 4 451 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000878.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pogz

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic sister chromatid cohesion
cell division
kinetochore assembly
nucleic acid binding
DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006730.mm.1 NM_001165948.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006730.mm.1 NM_001165948.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006730.mm.1 NM_172683.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006730.mm.1 NM_172683.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300006730.mm.1 TC0300000878.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0