Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200010873.mm.1 |
NM_001080754.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010873.mm.1 |
NM_001080754.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010873.mm.1 |
NM_172669.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010873.mm.1 |
NM_172669.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010873.mm.1 |
TC0200001382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010872.mm.1 |
NM_001080754.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010872.mm.1 |
NM_001080754.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010872.mm.1 |
NM_172669.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010872.mm.1 |
NM_172669.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010872.mm.1 |
TC0200001382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010875.mm.1 |
NM_001080754.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010875.mm.1 |
NM_001080754.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010875.mm.1 |
NM_172669.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010875.mm.1 |
NM_172669.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010875.mm.1 |
TC0200001382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010870.mm.1 |
NM_001080754.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010870.mm.1 |
NM_001080754.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010870.mm.1 |
NM_172669.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010870.mm.1 |
NM_172669.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010870.mm.1 |
TC0200001382.mm.1 |
MTA TC |