This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00554088_m1
See other DARS2 GE Assays ›
Gene Symbol:
Dars2
Gene Name:
aspartyl-tRNA synthetase 2 (mitochondrial)
Gene Aliases:
5830468K18Rik
Chromosome Location:
Chr.1: 161040612 - 161070658 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00554088_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dars2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172644.3 NP_766232.1 9 - 10 1348 70
GenBank mRNA AK030952.1 - 9 - 10 1337 70
AK033634.1 - 8 - 9 2171 70
AK037337.1 - 9 - 10 1367 70
AK048454.1 - 9 - 10 1325 70
AK138809.1 - 9 - 10 1349 70
AK154133.1 - 9 - 10 1376 70
AK154348.1 - 9 - 10 1376 70
AK164885.1 - 9 - 10 1318 70
BC046429.1 - 2 - 3 113 70
BC057943.1 - 9 - 10 1372 70
BC076606.1 - 9 - 10 1340 30625515 70
BC089191.1 - 9 - 10 1322 30473852 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003371.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dars2

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
tRNA aminoacylation
nucleotide binding
nucleic acid binding
aminoacyl-tRNA ligase activity
aspartate-tRNA ligase activity
ATP binding
ligase activity
protein homodimerization activity
aspartate-tRNA(Asn) ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100027554.mm.1 NM_172644.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100027554.mm.1 TC0100003371.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0