This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00553327_m1
See other PAPOLG GE Assays ›
Gene Symbol:
Papolg
Gene Name:
poly(A) polymerase gamma
Gene Aliases:
9630006B20Rik
Chromosome Location:
Chr.11: 23860145 - 23895296 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00553327_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Papolg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172555.2 NP_766143.2 15 - 16 1630 122
GenBank mRNA AK035810.1 - 15 - 16 1613 122
BC049163.1 - 15 - 16 1576 122
BC059271.1 - 15 - 16 1624 2811338 122
DQ103505.1 - 15 - 16 1436 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002379.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Papolg

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
RNA 3'-end processing
RNA polyadenylation
nucleotide binding
RNA binding
polynucleotide adenylyltransferase activity
ATP binding
transferase activity
nucleotidyltransferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100022416.mm.1 NM_172555.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100022416.mm.1 NM_172555.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100022416.mm.1 TC1100002379.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0