This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552439_g1
See other NUP88 GE Assays ›
Gene Symbol:
Nup88
Gene Name:
nucleoporin 88
Gene Aliases:
Nup84, Prei2
Chromosome Location:
Chr.11: 70943058 - 70969973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552439_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nup88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083331.2 NP_001076800.1 7 - 8 1219 67
NM_001276406.1 NP_001263335.1 7 - 8 1219 67
NM_172394.3 NP_765982.2 7 - 8 1219 67
GenBank mRNA AK002643.1 - 7 - 8 1205 67
AK028563.1 - 7 - 8 1209 67
AK039007.1 - 7 - 8 1209 67
AK050893.1 - 7 - 8 1208 67
AK085776.1 - 7 - 8 1218 67
AK160619.1 - 7 - 8 1220 67
AK166214.1 - 7 - 8 1209 67
BC032929.1 - 7 - 8 1200 1379673 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003117.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nup88

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s)

transport
protein transport
mRNA transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100028831.mm.1 NM_001083331.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100028831.mm.1 NM_001276406.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100028831.mm.1 NM_172394.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100028831.mm.1 TC1100003117.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0