This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552426_m1
See other AHSA2 GE Assays ›
Gene Symbol:
Ahsa2
Gene Name:
AHA1, activator of heat shock protein ATPase 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 23487882 - 23498039 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552426_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ahsa2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290654.1 NP_001277583.1 2 - 3 455 90
NM_001290655.1 NP_001277584.1 2 - 3 460 90
NM_172391.4 NP_765979.3 2 - 3 460 90
GenBank mRNA AK033696.1 - 2 - 3 368 90
AK035941.1 - 2 - 3 233 90
AK093945.1 - 2 - 3 367 90
AK137116.1 - 2 - 3 371 90
AK154196.1 - 2 - 3 371 90
AK155265.1 - 2 - 3 371 90
BC038397.1 - 2 - 3 346 3676293 90
BC052829.1 - 2 - 3 378 30040034 90
BC055873.1 - 2 - 3 322 90
BC064012.1 - 2 - 3 280 30135289 90
BY023007.1 - 2 - 3 345 90
CJ147821.1 - 2 - 3 461 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002364.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC038397.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ahsa2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

response to stress
biological_process
ATPase activator activity
molecular_function
chaperone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100022284.mm.1 NM_172391.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100022284.mm.1 TC1100002364.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100022285.mm.1 NM_172391.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100022285.mm.1 TC1100002364.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0