This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552314_g1
See other CPSF7 GE Assays ›
Gene Symbol:
Cpsf7
Gene Name:
cleavage and polyadenylation specific factor 7
Gene Aliases:
5730453I16Rik, AL022757, C330017N18Rik
Chromosome Location:
Chr.19: 10525244 - 10547735 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552314_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164272.1 NP_001157744.1 4 - 5 521 62
NM_172302.3 NP_758506.3 4 - 5 521 62
GenBank mRNA AK044306.1 - 4 - 5 501 62
AK053375.1 - 5 - 6 566 62
AK088603.1 - 4 - 5 499 62
AK165330.1 - 4 - 5 499 62
AK184839.1 - 2 - 3 151 62
BC017519.1 - 3 - 4 441 62
BC038812.1 - 3 - 4 451 4506179 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000201.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK053375.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf7

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
protein tetramerization
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002527.mm.1 NM_001164272.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002527.mm.1 NM_001164272.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002527.mm.1 NM_172302.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002527.mm.1 NM_172302.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900002527.mm.1 TC1900000201.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0