This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552041_m1
See other SLC24A4 GE Assays ›
Gene Symbol:
Slc24a4
Gene Name:
solute carrier family 24 (sodium/potassium/calcium exchanger), member 4
Gene Aliases:
A930002M03Rik, AW492432, NCKX4
Chromosome Location:
Chr.12: 102128652 - 102269083 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552041_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc24a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172152.2 NP_742164.1 2 - 3 212 81
GenBank mRNA AK044239.1 - 2 - 3 901 81
AK044368.1 - 2 - 3 905 81
AY156046.1 - 2 - 3 212 81
BC137945.1 - 2 - 3 240 8860964 81
BC171939.1 - 2 - 3 240 9054225 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001010.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc24a4

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
sodium ion transport
calcium ion transport
sensory perception of smell
sodium ion transmembrane transport
response to stimulus
transmembrane transport
calcium ion transmembrane transport
potassium ion transmembrane transport
amelogenesis
calcium:sodium antiporter activity
calcium, potassium:sodium antiporter activity
symporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200007099.mm.1 NM_172152.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007099.mm.1 NM_172152.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200007099.mm.1 TC1200001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200007100.mm.1 NM_172152.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007100.mm.1 NM_172152.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200007100.mm.1 TC1200001010.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200007098.mm.1 NM_172152.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007098.mm.1 NM_172152.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200007098.mm.1 TC1200001010.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0