This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00551234_m1
See other TDP1 GE Assays ›
Gene Symbol:
Tdp1
Gene Name:
tyrosyl-DNA phosphodiesterase 1
Gene Aliases:
2810481F14Rik, 4921509N21Rik, AI838772, AW493413, E430034L06Rik, MLZ-501, SCAN1
Chromosome Location:
Chr.12: 99884515 - 99955219 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00551234_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tdp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028354.4 NP_082630.2 11 - 12 1412 65
GenBank mRNA AK013427.1 - 10 - 11 887 65
AK014855.1 - 11 - 12 1392 65
AK033868.1 - 11 - 12 1410 65
BC019804.1 - 8 - 9 736 4018534 65
BC065162.1 - 10 - 11 1382 6402921 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000988.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tdp1

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

single strand break repair
DNA repair
double-strand break repair
cellular response to DNA damage stimulus
double-stranded DNA binding
single-stranded DNA binding
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
nuclease activity
exonuclease activity
phosphoric diester hydrolase activity
hydrolase activity
3'-tyrosyl-DNA phosphodiesterase activity
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200006882.mm.1 NM_028354.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006882.mm.1 NM_028354.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200006882.mm.1 TC1200000988.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006886.mm.1 NM_028354.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006886.mm.1 NM_028354.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200006886.mm.1 TC1200000988.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006885.mm.1 NM_028354.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006885.mm.1 NM_028354.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200006885.mm.1 TC1200000988.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0