This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00551225_m1
See other TDP1 GE Assays ›
Gene Symbol:
Tdp1
Gene Name:
tyrosyl-DNA phosphodiesterase 1
Gene Aliases:
2810481F14Rik, 4921509N21Rik, AI838772, AW493413, E430034L06Rik, MLZ-501, SCAN1
Chromosome Location:
Chr.12: 99884515 - 99955219 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00551225_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tdp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028354.4 NP_082630.2 2 - 3 603 114
GenBank mRNA AK033868.1 - 2 - 3 601 114
BC065162.1 - 1 - 2 573 6402921 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tdp1

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

single strand break repair
DNA repair
double-strand break repair
cellular response to DNA damage stimulus
double-stranded DNA binding
single-stranded DNA binding
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
nuclease activity
exonuclease activity
phosphoric diester hydrolase activity
hydrolase activity
3'-tyrosyl-DNA phosphodiesterase activity
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 531780 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0