Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400006981.mm.1 |
NM_027436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006981.mm.1 |
NM_027436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006981.mm.1 |
NR_040642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006981.mm.1 |
NR_040642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006981.mm.1 |
TC1400000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006977.mm.1 |
NM_027436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006977.mm.1 |
NM_027436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006977.mm.1 |
NR_040642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006977.mm.1 |
NR_040642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006977.mm.1 |
TC1400000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006978.mm.1 |
NM_027436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006978.mm.1 |
NM_027436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006978.mm.1 |
NR_040642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006978.mm.1 |
NR_040642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006978.mm.1 |
TC1400000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006979.mm.1 |
NM_027436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006979.mm.1 |
NM_027436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006979.mm.1 |
NR_040642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006979.mm.1 |
NR_040642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006979.mm.1 |
TC1400000854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006980.mm.1 |
NM_027436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006980.mm.1 |
NM_027436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006980.mm.1 |
NR_040642.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006980.mm.1 |
NR_040642.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006980.mm.1 |
TC1400000854.mm.1 |
MTA TC |