This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550855_g1
See other BAP1 GE Assays ›
Gene Symbol:
Bap1
Gene Name:
Brca1 associated protein 1
Gene Aliases:
2300006C11Rik, AA989761, AW553466, mKIAA0272, uch-x4
Chromosome Location:
Chr.14: 31251455 - 31259930 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550855_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027088.2 NP_081364.1 13 - 14 1861 82
GenBank mRNA AB047820.1 - 13 - 14 1762 82
AK079374.1 - 13 - 14 1879 82
AK129108.1 - 13 - 14 1769 82
AK170576.1 - 13 - 14 1862 82
AK171561.1 - 13 - 14 1864 82
BC042489.1 - 12 - 13 2309 82
BC050901.1 - 13 - 14 1824 6402443 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000326.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bap1

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
proteolysis
ubiquitin-dependent protein catabolic process
negative regulation of cell proliferation
chromatin modification
protein deubiquitination
monoubiquitinated protein deubiquitination
monoubiquitinated histone H2A deubiquitination
regulation of cell cycle
protein K48-linked deubiquitination
positive regulation of protein targeting to mitochondrion
chromatin binding
thiol-dependent ubiquitin-specific protease activity
protein binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400002823.mm.1 NM_027088.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002823.mm.1 NM_027088.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400002823.mm.1 TC1400000326.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0