This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00550557_m1
See other DIAP2 GE Assays ›
Gene Symbol:
Diap2
Gene Name:
diaphanous homolog 2 (Drosophila)
Gene Aliases:
Dia3, Diaph2, Drf2, E430022I22Rik
Chromosome Location:
Chr.X: 129749668 - 130465834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00550557_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Diap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172493.2 NP_766081.1 19 - 20 2482 73
GenBank mRNA AB713187.1 - 20 - 21 2277 73
AK083254.1 - 12 - 13 1414 73
AK088648.1 - 19 - 20 2416 73
AK153986.1 - 19 - 20 2470 73
AK197813.1 - 1 - 2 73 73
AY312280.1 - 19 - 20 2419 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001254.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
multicellular organism development
female gamete generation
cellular component organization
actin cytoskeleton organization
actin filament polymerization
cell differentiation
oogenesis
actin binding
Rho GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00007779.mm.1 NM_172493.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007779.mm.1 NM_172493.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007779.mm.1 TC0X00001254.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0