This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00549996_m1
See other NLRP4B GE Assays ›
Gene Symbol:
Nlrp4b
Gene Name:
NLR family, pyrin domain containing 4B
Gene Aliases:
AU022726, E330024M09, Nalp-gamma, Nalp4b
Chromosome Location:
Chr.7: 10687793 - 10730168 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00549996_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nlrp4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172481.2 NP_766069.1 8 - 9 2669 69
GenBank mRNA AK054426.1 - 8 - 9 2670 69
AY355342.1 - 8 - 9 2670 69
AY596198.1 - 6 - 7 2505 69
BC066795.1 - 8 - 9 2679 30473929 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000110.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nlrp4b

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
biological_process
nucleotide binding
molecular_function
ATP binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011902 BC066795.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011902 AY355342.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011902 NM_172481.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011902 AK054426.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519698 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336331 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961747 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199389 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 AF119253.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 AF119254.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 AK002532.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 BC019721.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 BC031711.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 K01922.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 K01926.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 M11357.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 M21931.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 NM_010378.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 V00832.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431279 X52643.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820876 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431947 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695490 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685560 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83723 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264537 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155073 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946846 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493697 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751630 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916654 AK017250.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916654 AK017424.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916654 BC087927.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916654 NM_009664.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916654 U65021.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685802 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106798 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86517 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168313 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147988 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103045 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011902 BC066795.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011902 AY355342.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011902 NM_172481.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011902 AK054426.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966932 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966952 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966939 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966938 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966950 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966961 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966931 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966969 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966929 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966960 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966963 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966951 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966948 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966927 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966935 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966956 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966968 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966962 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966941 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966943 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966946 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966936 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966949 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966965 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966947 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06122507 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06122522 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06122463 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147988 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01478590 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103045 - -
Whole Mouse Genome G4122A A_51_P287790 BC066795.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P287790 AY355342.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P287790 NM_172481.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P287790 AK054426.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P572679 BC066795.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P572679 AY355342.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P572679 NM_172481.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P572679 AK054426.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134543 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135052 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103045 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147988 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06517384 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06446172 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06446166 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06638581 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06446163 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06584818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967067 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967051 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966967 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966943 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967101 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966935 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967053 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966989 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966980 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967087 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966993 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966962 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966968 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967081 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966960 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967036 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967116 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967119 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967077 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966997 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966951 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966975 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967122 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966948 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24967143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966990 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966935 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966943 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966970 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966950 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966956 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966938 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966941 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06446172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966975 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966960 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06639758 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966946 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966978 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05933592 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966968 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06446170 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966928 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966961 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966967 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966984 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966927 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966963 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966977 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966965 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966969 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966944 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966929 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966926 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06584819 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06463327 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966987 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966948 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966980 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966983 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966942 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24966951 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10549956 - -
Affymetrix Mouse Gene 1.0 ST Array 10549961 - -
Affymetrix Mouse Gene 1.0 ST Array 10549957 - -
Affymetrix Mouse Gene 1.0 ST Array 10549950 - -
Affymetrix Mouse Gene 1.0 ST Array 10549955 - -
Affymetrix Mouse Gene 1.0 ST Array 10549958 - -
Affymetrix Mouse Gene 1.0 ST Array 10549959 - -
Affymetrix Mouse Gene 1.0 ST Array 10549954 - -
Affymetrix Mouse Gene 1.0 ST Array 10549953 - -
Affymetrix Mouse Gene 1.0 ST Array 10549952 - -
Affymetrix Mouse Gene 1.0 ST Array 10549960 - -
Affymetrix HT MG-430 PM Array Plate 1434527_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10549949 - -
Mouse Genome 430 2.0 Array 1434527_at - -
Mouse Genome 430 2.0 Array 1434527_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 210045_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 210045_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700001001.mm.1 NM_172481.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001001.mm.1 NM_172481.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700001001.mm.1 TC0700000110.mm.1 MTA TC
Mouse Expression Set 430 1434527_PM_at - -
Mouse Expression Set 430 1434527_at - -
Murine Genome U74v2 Set 135521_PM_r_at - -
Murine Genome U74v2 Set 135521_r_at - -
Murine Genome U74v2 Set 135520_PM_f_at - -
Murine Genome U74v2 Set 135520_f_at - -
Affymetrix HT Mouse Genome 430B Array 1434527_at - -
Affymetrix HT Mouse Genome 430B Array 1434527_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 824861 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2060075 - -
Illumina mouse-6 v1.0 expression beadchip 2060075 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002060075 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649484 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2649484 - -
mouse-6 v1.1 (Illumina) scl33086.11.1_97-S - -
Illumina Sentrix mouse-6.v1 bead array 2060075 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl33086.11.1_97-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060075 - -
MouseRef-8 v2 Expression BeadChip ILMN_2649484 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2649484 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0