This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00545933_m1
See other CRYGA GE Assays ›
Gene Symbol:
Cryga
Gene Name:
crystallin, gamma A
Gene Aliases:
Cryg-4, DGcry-4
Chromosome Location:
Chr.1: 65100389 - 65103363 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00545933_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cryga
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007774.3 NP_031800.1 2 - 3 280 145
GenBank mRNA BC056430.1 - 2 - 3 267 6824353 145
BY150722.1 - 2 - 3 291 145
CF728682.1 - 2 - 3 280 145
CF738355.1 - 2 - 3 280 145

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cryga

Gene Ontology Categories:

Function(s) Process(es)

eye development
lens development in camera-type eye
structural constituent of eye lens

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF738355.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 NM_007774.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 BC056430.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF728682.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF738355.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 NM_007774.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 BC056430.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF728682.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639989 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115334 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF738355.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 NM_007774.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 BC056430.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF728682.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF738355.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 NM_007774.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 BC056430.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF728682.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF738355.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 NM_007774.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 BC056430.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF728682.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF738355.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 NM_007774.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 BC056430.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF728682.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296427 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296388 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296390 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296405 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296403 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296425 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296406 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296396 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296421 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296412 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296423 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296416 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296414 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296408 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296420 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296392 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296393 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296394 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296410 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296417 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115334 - -
Whole Mouse Genome G4122A A_51_P508156 CF738355.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 NM_007774.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 BC056430.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 CF728682.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 CF738355.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 NM_007774.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 BC056430.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P508156 CF728682.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116357 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115334 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00015179 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00156850 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00015184 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00015185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296417 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296364 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00015179 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296405 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296414 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296424 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296385 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296393 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296410 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296426 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296400 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296381 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296379 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296377 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00781477 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296416 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00156850 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296392 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296390 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296386 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296382 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296395 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00015181 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00156849 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00156847 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296406 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296423 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20296389 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10355204 - -
Affymetrix Mouse Gene 1.0 ST Array 10355203 - -
Affymetrix HT MG-430 PM Array Plate 1422652_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454333_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10355202 - -
Mouse Genome 430 2.0 Array 1454333_at - -
Mouse Genome 430 2.0 Array 1422652_PM_at - -
Mouse Genome 430 2.0 Array 1454333_PM_at - -
Mouse Genome 430 2.0 Array 1422652_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12964_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12964_at - -
Mouse Expression Set 430 1422652_PM_at - -
Mouse Expression Set 430 1422652_at - -
Mouse Expression Set 430 1454333_PM_at - -
Mouse Expression Set 430 1454333_at - -
Murine Genome U74v2 Set 103609_at - -
Murine Genome U74v2 Set 103609_PM_at - -
Murine Genome U74v2 Set 169050_PM_r_at - -
Murine Genome U74v2 Set 169050_r_at - -
Mouse Genome 430A 2.0 Array 1422652_at - -
Mouse Genome 430A 2.0 Array 1422652_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1422652_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1422652_at - -
Affymetrix HT Mouse Genome 430B Array 1454333_at - -
Affymetrix HT Mouse Genome 430B Array 1454333_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 850560 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2340309 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002340309 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705692 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2705692 - -
mouse-6 v1.1 (Illumina) scl012964.1_261-S - -
Illumina Sentrix mouse-6.v1 bead array 2340309 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl012964.1_261-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340309 - -
MouseRef-8 v2 Expression BeadChip ILMN_2705692 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2705692 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0