This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00525545_m1
See other LRIT1 GE Assays ›
Gene Symbol:
Lrit1
Gene Name:
leucine-rich repeat, immunoglobulin-like and transmembrane domains 1
Gene Aliases:
BC032270, Lrrc21
Chromosome Location:
Chr.14: 37054830 - 37064938 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00525545_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lrit1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146245.2 NP_666357.2 3 - 4 992 66
GenBank mRNA AK044631.1 - 3 - 4 993 66
BC032270.1 - 3 - 4 998 5401567 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000392.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lrit1

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979295 NM_146245.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979295 AK044631.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979295 BC032270.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958258 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510253 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401164 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176923 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89649 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143097 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168790 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173717 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979295 NM_146245.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979295 AK044631.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979295 BC032270.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542960 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543006 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543012 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542965 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543008 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543011 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542971 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543020 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543014 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543016 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542953 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542958 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542956 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542966 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542968 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543018 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542961 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29542963 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543010 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29543017 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02834838 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173717 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168790 - -
Whole Mouse Genome G4122A A_51_P145020 NM_146245.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P145020 AK044631.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P145020 BC032270.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132225 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173717 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168790 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11737591 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11737594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543020 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543011 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542939 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543064 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542966 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543043 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542951 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542991 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542999 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543049 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542953 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542946 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543028 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542968 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543087 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542985 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542977 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542986 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542989 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542907 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542997 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543073 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542995 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542993 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29543068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29542908 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11661940 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542965 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11737590 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11737581 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543003 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11652179 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542961 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542997 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542956 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11292112 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543008 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542993 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542974 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542946 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542994 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543014 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11661947 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542958 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542977 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11702057 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542976 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542963 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542992 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543007 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542991 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542960 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11168751 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543012 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543018 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542939 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542980 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542973 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11702059 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11292101 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11292106 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542954 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543016 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542951 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543004 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543020 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11702072 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11292104 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11661948 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542950 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542966 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11737580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543009 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542941 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29542988 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11737591 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11409070 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29543011 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10414164 - -
Affymetrix Mouse Gene 1.0 ST Array 10414167 - -
Affymetrix Mouse Gene 1.0 ST Array 10414166 - -
Affymetrix Mouse Gene 1.0 ST Array 10414165 - -
Affymetrix HT MG-430 PM Array Plate 1441110_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10414163 - -
Mouse Genome 430 2.0 Array 1441110_PM_at - -
Mouse Genome 430 2.0 Array 1441110_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 239037_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 239037_at - -
Affymetrix Mouse Clariom S Array PSR1400003491.mm.1 NM_146245.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003491.mm.1 NM_146245.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400003491.mm.1 TC1400000392.mm.1 MTA TC
Mouse Expression Set 430 1441110_PM_at - -
Mouse Expression Set 430 1441110_at - -
Affymetrix HT Mouse Genome 430B Array 1441110_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1441110_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 460676 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 7040736 - -
Illumina mouseRef-8 v1.0 expression beadchip 0007040736 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589757 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2589757 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2525924 - -
mouse-6 v1.1 (Illumina) scl46440.4.1_161-S - -
Illumina Sentrix mouse-6.v1 bead array 7040736 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl46440.4.1_161-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040736 - -
MouseRef-8 v2 Expression BeadChip ILMN_2589757 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589757 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0