This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00525322_m1
See other C87436 GE Assays ›
Gene Symbol:
C87436
Gene Name:
expressed sequence C87436
Gene Aliases:
-
Chromosome Location:
Chr.6: 86437328 - 86470388 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00525322_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C87436
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243742.1 NP_001230671.1 3 - 4 1229 103
NM_146170.4 NP_666282.3 3 - 4 1170 103
GenBank mRNA AK047127.1 - 3 - 4 1171 103
AK077101.1 - 996 103
AK170524.1 - 4 - 5 1061 103
AK171511.1 - 4 - 5 1061 103
AK178467.1 - 1 - 2 159 103
BC025632.1 - 3 - 4 1137 5321669 103
BC026926.1 - 3 - 4 1240 3586097 103
BC031913.1 - 3 - 4 1183 5376246 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000989.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene C87436

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600007704.mm.1 NM_001243742.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007704.mm.1 NM_001243742.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007704.mm.1 NM_146170.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007704.mm.1 NM_146170.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007704.mm.1 TC0600000989.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header