This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523308_m1
See other PTBP3 GE Assays ›
Gene Symbol:
Ptbp3
Gene Name:
polypyrimidine tract binding protein 3
Gene Aliases:
5830471K22Rik, AA407443, AI462022, AW107884, C86549, Rod1
Chromosome Location:
Chr.4: 59471868 - 59549379 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523308_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptbp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144904.2 NP_659153.2 9 - 10 1169 62
NM_178164.3 NP_835458.1 8 - 9 1144 62
GenBank mRNA AK033732.1 - 8 - 9 1069 62
AK042920.1 - 6 - 7 1076 62
AK088560.1 - 8 - 9 1076 62
BC006638.1 - 9 - 10 1079 3969488 62
BC057641.1 - 8 - 9 1049 4936557 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002757.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptbp3

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
cell differentiation
negative regulation of RNA splicing
erythrocyte maturation
regulation of cell differentiation
negative regulation of mRNA splicing, via spliceosome
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400022368.mm.1 NM_144904.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022368.mm.1 NM_144904.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400022368.mm.1 NM_178164.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022368.mm.1 NM_178164.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400022368.mm.1 TC0400002757.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0