This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523306_m1
See other PTBP3 GE Assays ›
Gene Symbol:
Ptbp3
Gene Name:
polypyrimidine tract binding protein 3
Gene Aliases:
5830471K22Rik, AA407443, AI462022, AW107884, C86549, Rod1
Chromosome Location:
Chr.4: 59471868 - 59549379 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523306_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptbp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144904.2 NP_659153.2 7 - 8 921 63
NM_178164.3 NP_835458.1 6 - 7 896 63
GenBank mRNA AK033732.1 - 6 - 7 821 63
AK042920.1 - 4 - 5 828 63
AK088560.1 - 6 - 7 828 63
BC006638.1 - 7 - 8 831 3969488 63
BC057641.1 - 6 - 7 801 4936557 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002757.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptbp3

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
cell differentiation
negative regulation of RNA splicing
erythrocyte maturation
regulation of cell differentiation
negative regulation of mRNA splicing, via spliceosome
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400022374.mm.1 NM_144904.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022374.mm.1 NM_144904.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400022374.mm.1 NM_178164.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022374.mm.1 NM_178164.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400022374.mm.1 TC0400002757.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400022375.mm.1 NM_144904.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022375.mm.1 NM_144904.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400022375.mm.1 NM_178164.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022375.mm.1 NM_178164.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400022375.mm.1 TC0400002757.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0