This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523119_m1
See other VASH2 GE Assays ›
Gene Symbol:
Vash2
Gene Name:
vasohibin 2
Gene Aliases:
B130052G07Rik
Chromosome Location:
Chr.1: 190947646 - 190979303 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523119_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vash2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144879.2 NP_659128.2 3 - 4 796 78
NR_027352.1 3 - 4 663 78
GenBank mRNA AK045261.1 - 3 - 4 592 78
AK078596.1 - 3 - 4 665 78
AK140989.1 - 3 - 4 790 78
AK163193.1 - 3 - 4 572 78
BC024141.1 - 3 - 4 566 5149750 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vash2

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endothelial cell proliferation
regulation of angiogenesis
positive regulation of angiogenesis
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0