This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522876_m1
See other ICE1 GE Assays ›
Gene Symbol:
Ice1
Gene Name:
interactor of little elongation complex ELL subunit 1
Gene Aliases:
C77245, mKIAA0947
Chromosome Location:
Chr.13: 70588688 - 70637838 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522876_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ice1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144837.3 NP_659086.2 15 - 16 6155 97
GenBank mRNA AK129244.1 - 3 - 4 4592 97
AK134564.1 - 3 - 4 2598 97
BC018507.1 - 2 - 3 250 97
BC026613.1 - 3 - 4 681 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002285.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ice1

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of protein complex assembly
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
positive regulation of transcription from RNA polymerase III promoter
positive regulation of intracellular protein transport
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 BC026613.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 AK129244.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 AK134564.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 BC018507.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 NM_144837.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 BC026613.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 AK129244.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 AK134564.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 BC018507.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232439 NM_144837.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941489 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155791 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777826 AK004980.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777826 AK077968.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777826 AK163403.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777826 NM_008615.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777826 XM_001004685.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298395 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238622 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839289 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922701 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867488 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582725 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654694 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895782 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956680 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787755 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53978 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789619 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948500 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48569 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33291 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108093 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145550 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101849 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161147 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 BC026613.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 AK129244.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 AK134564.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 BC018507.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 NM_144837.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 BC026613.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 AK129244.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 AK134564.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 BC018507.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232439 NM_144837.3 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07911424 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07911466 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07911564 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02715531 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101849 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02715519 - -
Whole Mouse Genome G4122A A_51_P232439 BC026613.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 AK129244.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 AK134564.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 BC018507.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 NM_144837.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 BC026613.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 AK129244.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 AK134564.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 BC018507.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P232439 NM_144837.3 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101849 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07911564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122454 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10963789 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122656 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122592 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122442 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10628988 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122425 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122463 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122441 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122398 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122447 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122492 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585821 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122591 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122457 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122436 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10676904 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122373 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122596 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585741 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122544 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122483 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10628969 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122429 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122587 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122420 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122481 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122384 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585702 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122473 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122423 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122598 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122468 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122658 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122484 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122370 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585714 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122402 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122602 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122400 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122495 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122385 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122646 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122478 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10628995 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585829 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122609 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122550 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122459 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122432 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122641 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122667 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585775 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122595 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122662 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122387 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122546 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122450 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122412 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122664 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122674 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122480 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10691172 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122401 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122461 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10676900 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122488 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122616 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122582 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122405 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122599 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122379 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122539 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122547 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122589 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10640540 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122553 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122383 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122491 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122618 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122409 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585754 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122411 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122389 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122493 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122610 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122601 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122448 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122395 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122653 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122382 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122417 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122661 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585757 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122476 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122594 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122597 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585793 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10726222 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122581 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122644 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122426 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122486 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122367 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585725 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585773 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122456 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122489 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10964201 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585711 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10828848 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10726221 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122399 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10636576 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122464 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122621 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122666 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122368 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10628971 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122628 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585796 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122603 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122654 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10585762 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10828856 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29122499 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10410470 - -
Affymetrix Mouse Gene 1.0 ST Array 10410468 - -
Affymetrix Mouse Gene 1.0 ST Array 10410469 - -
Affymetrix Mouse Gene 1.0 ST Array 10410472 - -
Affymetrix Mouse Gene 1.0 ST Array 10410466 - -
Affymetrix Mouse Gene 1.0 ST Array 10410471 - -
Affymetrix Mouse Gene 1.0 ST Array 10410474 - -
Affymetrix Mouse Gene 1.0 ST Array 10410473 - -
Affymetrix Mouse Gene 1.0 ST Array 10410467 - -
Affymetrix HT MG-430 PM Array Plate 1424071_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1451221_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10410465 - -
Mouse Genome 430 2.0 Array 1451221_at - -
Mouse Genome 430 2.0 Array 1451221_PM_at - -
Mouse Genome 430 2.0 Array 1424071_s_at - -
Mouse Genome 430 2.0 Array 1424071_PM_s_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 218333_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 218333_at - -
Affymetrix Mouse Clariom S Array PSR1300014931.mm.1 NM_144837.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014931.mm.1 NM_144837.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300014931.mm.1 TC1300002285.mm.1 MTA TC
Mouse Expression Set 430 1424071_s_at - -
Mouse Expression Set 430 1451221_at - -
Mouse Expression Set 430 1424071_PM_s_at - -
Mouse Expression Set 430 1451221_PM_at - -
Murine Genome U74v2 Set 94979_PM_at - -
Murine Genome U74v2 Set 94979_at - -
Mouse Genome 430A 2.0 Array 1424071_PM_s_at - -
Mouse Genome 430A 2.0 Array 1451221_at - -
Mouse Genome 430A 2.0 Array 1451221_PM_at - -
Mouse Genome 430A 2.0 Array 1424071_s_at - -
Affymetrix HT Mouse Genome 430A Array 1424071_s_at - -
Affymetrix HT Mouse Genome 430A Array 1451221_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1424071_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1451221_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 524666 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5340324 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259805 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230422 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1259805 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1230422 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0