This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00522102_g1
See other MBD1 GE Assays ›
Gene Symbol:
Mbd1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
Cxxc3, PCM1
Chromosome Location:
Chr.18: 74267615 - 74282734 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00522102_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013594.2 NP_038622.2 4 - 5 646 78
GenBank mRNA AF072240.1 - 5 - 6 625 78
AK004624.2 - 4 - 5 647 78
AK032535.1 - 1 - 2 144 78
AK156401.1 - 5 - 6 637 78
AK162858.1 - 4 - 5 646 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000788.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbd1

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of gene expression
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800005655.mm.1 NM_013594.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005655.mm.1 NM_013594.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800005655.mm.1 TC1800000788.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0